siRNA Details
| Gene Symbol: Dok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107336.1 | NP_001100806.1 | 5 | 1203 | ||
| XM_006253619.2 | XP_006253681.1 | 5 | 1215 | |||
| GenBank mRNA | BC166772.1 | - | 5 | 1195 | 7934143 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok3| Gene Symbol: Dok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107336.1 | NP_001100806.1 | 5 | 1203 | ||
| XM_006253619.2 | XP_006253681.1 | 5 | 1215 | |||
| GenBank mRNA | BC166772.1 | - | 5 | 1195 | 7934143 | |