siRNA Details
| Gene Symbol: Mov10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107711.1 | NP_001101181.1 | 11, 12 | 2049 | ||
| XM_006233085.1 | XP_006233147.1 | 11, 12 | 1937 | |||
| XM_008761395.1 | XP_008759617.1 | 11, 12 | 2797 | |||

Mov10
Genomic Map