siRNA Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108016.2 | NP_001101486.2 | 4 | 464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mboat2| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108016.2 | NP_001101486.2 | 4 | 464 | ||