This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PCNX SIRNA Assays ›
Gene Symbol:
Pcnx
Gene Name:
pecanex homolog (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.6: 105611430 - 105747176 on Build Rnor_6.0
UniGene ID:
Rn.23381
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359269
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pcnx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170347.1 NP_001163818.1 6 905
XM_006240299.2 XP_006240361.1 6 931
XM_006240300.2 XP_006240362.1 6 931
XM_006240301.1 XP_006240363.1 7 967
XM_006240302.2 XP_006240364.1 6 931
XM_008764788.1 XP_008763010.1 5 841
XM_008764789.1 XP_008763011.1 6 2327

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline