siRNA Details
| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108232.2 | NP_001101702.1 | 1, 2 | 763 | ||
| GenBank mRNA | BC168978.1 | - | 1, 2 | 763 | 7455775 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb1| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108232.2 | NP_001101702.1 | 1, 2 | 763 | ||
| GenBank mRNA | BC168978.1 | - | 1, 2 | 763 | 7455775 | |