siRNA Details
| Gene Symbol: Dnajb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015021.1 | NP_001015021.1 | 9 | 1109 | ||
| GenBank mRNA | AY387070.1 | - | 9 | 929 | ||
| BC093384.1 | - | 9 | 1109 | 7389370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb11| Gene Symbol: Dnajb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015021.1 | NP_001015021.1 | 9 | 1109 | ||
| GenBank mRNA | AY387070.1 | - | 9 | 929 | ||
| BC093384.1 | - | 9 | 1109 | 7389370 | ||