This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SUCO SIRNA Assays ›
Gene Symbol:
Suco
Gene Name:
SUN domain containing ossification factor
Gene Aliases:
Dd25
Chromosome Location:
Chr.13: 79736485 - 79801574 on Build Rnor_6.0
UniGene ID:
Rn.228937
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359906
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Suco
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199403.1 NP_955435.1 18 2543
XM_006250164.2 XP_006250226.1 18 2998
XM_006250166.2 XP_006250228.1 17 2977
XM_008769657.1 XP_008767879.1 18 3010
XM_008769658.1 XP_008767880.1 17 2989
XM_008769659.1 XP_008767881.1 18 3010
GenBank mRNA AJ421447.1 - 18 2543

Back To Top

More Information


Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s)

ossification
multicellular organism development
positive regulation of collagen biosynthetic process
positive regulation of osteoblast differentiation
regulation of bone remodeling

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline