siRNA Details
Gene Symbol: Mrgprx2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002285.1 | NP_001002285.1 | 1 | 673 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
