siRNA Details
| Gene Symbol: LOC100364500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255892.2 | XP_006255954.1 | 1 | 120 | ||
| Gene Symbol: RT1-CE15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008838.1 | NP_001008838.1 | 2 | 483 | ||

LOC100364500
Genomic Map