siRNA Details
| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009536.1 | NP_001009536.1 | 8, 9 | 1462 | ||
| GenBank mRNA | BC061749.1 | - | 8, 9 | 1462 | 5599464 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim25| Gene Symbol: Trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009536.1 | NP_001009536.1 | 8, 9 | 1462 | ||
| GenBank mRNA | BC061749.1 | - | 8, 9 | 1462 | 5599464 | |