siRNA Details
| Gene Symbol: LOC498122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109058.1 | NP_001102528.1 | 3 | 236 | ||
| XM_008768900.1 | XP_008767122.1 | 2 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498122| Gene Symbol: LOC498122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109058.1 | NP_001102528.1 | 3 | 236 | ||
| XM_008768900.1 | XP_008767122.1 | 2 | 540 | |||