siRNA Details
Gene Symbol: Slamf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191932.1 | NP_001178861.1 | 2 | 217 | ||
XM_006250301.2 | XP_006250363.1 | 2 | 638 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slamf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191932.1 | NP_001178861.1 | 2 | 217 | ||
XM_006250301.2 | XP_006250363.1 | 2 | 638 |