siRNA Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126085.1 | NP_001119557.1 | 4, 5 | 408 | ||
| XM_006255408.1 | XP_006255470.1 | 3, 4 | 323 | |||
| Gene Symbol: LOC100360750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728002.4 | XP_002728048.1 | 1, 2 | 381 | ||

Lsm6
Genomic Map