This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM200A SIRNA Assays ›
Gene Symbol:
Tmem200a
Gene Name:
transmembrane protein 200A
Gene Aliases:
RGD1564024
Chromosome Location:
Chr.1: 20330667 - 20413921 on Build Rnor_6.0
UniGene ID:
Rn.162404
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS364140
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem200a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109135.1 NP_001102605.1 2 1762
XM_006227690.2 XP_006227752.1 3 3140
XM_008758623.1 XP_008756845.1 2 2690
XM_008758624.1 XP_008756846.1 3 1437
XM_008758625.1 XP_008756847.1 4 3245

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029881, AK165025, BC141267, AK173299, BC141266, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline