siRNA Details
| Gene Symbol: Olr765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234700.1 | XP_006234762.1 | 2 | 331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr765| Gene Symbol: Olr765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234700.1 | XP_006234762.1 | 2 | 331 | ||