siRNA Details
| Gene Symbol: Ppp1r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109222.1 | NP_001102692.1 | 3 | 961 | ||
| XM_008762717.1 | XP_008760939.1 | 3 | 1003 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r3a| Gene Symbol: Ppp1r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109222.1 | NP_001102692.1 | 3 | 961 | ||
| XM_008762717.1 | XP_008760939.1 | 3 | 1003 | |||