siRNA Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237697.2 | XP_006237759.1 | 5 | 1743 | ||
| XM_575726.6 | XP_575726.2 | 5 | 1040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Far2| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237697.2 | XP_006237759.1 | 5 | 1743 | ||
| XM_575726.6 | XP_575726.2 | 5 | 1040 | |||