This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GLI4 SIRNA Assays ›
Gene Symbol:
Gli4 LOC100911986
Gene Name:
GLI family zinc finger 4
zinc finger protein GLI4-like
Gene Aliases:
-
-
Chromosome Location:
Chr.7: 116671936 - 116677188 on Build Rnor_6.0
Chromosome Location:
Chr.7|NW_007906218: 28993 - 30948 on Build Rnor_6.0
UniGene ID:
Rn.18674
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS366059
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gli4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029926.1 NP_001025097.1 4 833
XM_006241856.2 XP_006241918.1 4 432
XM_006241858.1 XP_006241920.1 4 717
GenBank mRNA BC097383.1 - 4 833 7446306
Gene Symbol: LOC100911986
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752188.3 XP_003752236.2 1 550

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
nucleic acid binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline