siRNA Details
| Gene Symbol: Armcx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024367.1 | NP_001019538.1 | 632 | |||
| XM_006257253.2 | XP_006257315.1 | 4 | 1271 | |||
| GenBank mRNA | BC085780.1 | - | 632 | 7107145 | ||
| FQ221160.1 | - | 656 | ||||
| Gene Symbol: LOC102554790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257242.2 | XP_006257304.1 | 4 | 715 | ||
| XM_006257243.2 | XP_006257305.1 | 4 | 1271 | |||

Armcx1
Genomic Map