siRNA Details
| Gene Symbol: RGD1559662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767614.1 | XP_008765836.1 | 4 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559662| Gene Symbol: RGD1559662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767614.1 | XP_008765836.1 | 4 | 542 | ||