siRNA Details
| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205348.1 | NP_001192277.1 | 7 | 1220 | ||
| GenBank mRNA | EF199812.1 | - | 7 | 915 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd300a| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205348.1 | NP_001192277.1 | 7 | 1220 | ||
| GenBank mRNA | EF199812.1 | - | 7 | 915 | ||