This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC100912787 SIRNA Assays ›
Gene Symbol:
LOC100912787 LOC103690092
Gene Name:
zinc finger protein 850-like
zinc finger protein 790-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 88510260 - 88525397 on Build Rnor_6.0
Chromosome Location:
Chr.1: 87729490 - 87744456 on Build Rnor_6.0
UniGene ID:
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS366832
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100912787
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228762.2 XP_006228824.1 5 862
XM_006228763.2 XP_006228825.1 5 786
XM_006228764.2 XP_006228826.1 5 1222
XM_008759233.1 XP_008757455.1 5 873
Gene Symbol: LOC103690092
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759223.1 XP_008757445.1 786

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kgr6d:80/100.66.78.179:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline