siRNA Details
| Gene Symbol: Gpr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024925.2 | NP_001020096.1 | 3 | 889 | ||
| XM_006256675.2 | XP_006256737.1 | 4 | 3522 | |||
| XM_006256676.2 | XP_006256738.1 | 4 | 1439 | |||
| GenBank mRNA | AB159443.1 | - | 3 | 889 | ||
More Information
Other Species Targeted
Set Membership: |
|

Gpr34
Genomic Map