This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other POU5F2 SIRNA Assays ›
Gene Symbol:
Pou5f2
Gene Name:
POU domain class 5, transcription factor 2
Gene Aliases:
Sprm-1, Sprm1
Chromosome Location:
Chr.2: 5437067 - 5438301 on Build Rnor_6.0
UniGene ID:
Rn.109178
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS368959
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pou5f2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081751.1 NP_001075220.1 1 356
GenBank mRNA L23864.1 - 1 356

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
spermatogenesis
cell differentiation
meiotic cell cycle
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-cdc4cf686-5rvzh:80/100.66.79.50:80.
git-commit: 18f7f941bc72f70e8ff4e0be257f7363f2314623
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.0-Offline