siRNA Details
| Gene Symbol: LOC102549011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256982.2 | XP_006257044.1 | 2 | 1902 | ||
| XM_008773280.1 | XP_008771502.1 | 2 | 1902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102549011| Gene Symbol: LOC102549011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256982.2 | XP_006257044.1 | 2 | 1902 | ||
| XM_008773280.1 | XP_008771502.1 | 2 | 1902 | |||