This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESCO1 SIRNA Assays ›
Gene Symbol:
Esco1
Gene Name:
establishment of sister chromatid cohesion N-acetyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 1789796 - 1857269 on Build Rnor_6.0
UniGene ID:
Rn.19439
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS369524
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Esco1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126299.2 NP_001119771.2 1 1683
XM_006254412.2 XP_006254474.1 3 2296
XM_006254413.2 XP_006254475.1 3 2106
XM_006254414.2 XP_006254476.1 3 2293
XM_006254415.2 XP_006254477.1 3 2459
GenBank mRNA BC166441.1 - 1 1683 7461161

Back To Top

More Information


Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s)

regulation of DNA replication
sister chromatid cohesion
post-translational protein acetylation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0