This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP2K3 SIRNA Assays ›
Gene Symbol:
MAP2K3
Gene Name:
mitogen-activated protein kinase kinase 3
Gene Aliases:
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location:
Chr.17: 21284656 - 21315240 on Build GRCh38
UniGene ID:
Hs.514012
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID VHS40371
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP2K3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316332.1 NP_001303261.1 4, 5 543
NM_002756.4 NP_002747.2 3, 4 160
NM_145109.2 NP_659731.1 3, 4 418
XM_005256723.2 XP_005256780.1 4, 5 458
XM_011523958.2 XP_011522260.1 3, 4 217
XM_011523959.1 XP_011522261.1 5, 6 425
XM_017024857.1 XP_016880346.1 3, 4 287
XM_017024858.1 XP_016880347.1 4, 5 275
XM_017024859.1 XP_016880348.1 5, 6 615
GenBank mRNA AB451226.1 - 3, 4 153
AB451349.1 - 3, 4 153
AK093838.1 - 4, 5 462
AK223490.1 - 3, 4 293
AK315505.1 - 3, 4 363
BC032478.1 - 3, 4 353 5215093
BM836867.1 - 3, 4 398
CR536514.1 - 3, 4 153
CR541793.1 - 3, 4 153
D87116.1 - 3, 4 210
DA131175.1 - 3, 4 418
DA987047.1 - 3, 4 161
GQ225578.1 - 3, 4 153
L36719.1 - 5, 6 403
U66839.1 - 3, 4 279

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
inflammatory response
signal transduction
peptidyl-tyrosine phosphorylation
proteolysis in other organism
regulation of cytokine biosynthetic process
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
cardiac muscle contraction
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline