This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2410018L13RIK SIRNA Assays ›
Gene Symbol:
2410018L13Rik
Gene Name:
RIKEN cDNA 2410018L13 gene
Gene Aliases:
2810480C08Rik
Chromosome Location:
Chr.12: 22953997 - 23010243 on Build GRCm38
UniGene ID:
Mm.391403
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n251661

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2410018L13Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015504.1 4 838
GenBank mRNA AK134640.1 - 4 839
BC063063.1 - 5 2808

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-npfdm:80/100.66.76.105:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0