siRNA Details
| Gene Symbol: Emx2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002863.2 | 2 | 654 | |||
| GenBank mRNA | AK033011.1 | - | 2 | 303 | ||
| AY117414.1 | - | LIT1111 EMX2OS |
2 | 653 | ||

Emx2os
Genomic Map