siRNA Details
| Gene Symbol: 5031426D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027890.1 | 1 | 2523 | |||
| GenBank mRNA | AK019879.1 | - | 1 | 2522 | ||
| AK032294.1 | - | 1 | 711 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5031426D15Rik| Gene Symbol: 5031426D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027890.1 | 1 | 2523 | |||
| GenBank mRNA | AK019879.1 | - | 1 | 2522 | ||
| AK032294.1 | - | 1 | 711 | |||