siRNA Details
| Gene Symbol: 9230102K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028438.1 | 3 | 1252 | |||
| GenBank mRNA | AK078991.1 | - | 3 | 1254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230102K24Rik| Gene Symbol: 9230102K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028438.1 | 3 | 1252 | |||
| GenBank mRNA | AK078991.1 | - | 3 | 1254 | ||