siRNA Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027826.1 | 1 | 44 | |||
| GenBank mRNA | AK011437.1 | - | 1 | 44 | ||
| AK019154.1 | - | 1 | 44 | |||
| AK048630.1 | - | 1 | 42 | |||
| BC094545.1 | - | 24 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pantr1| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027826.1 | 1 | 44 | |||
| GenBank mRNA | AK011437.1 | - | 1 | 44 | ||
| AK019154.1 | - | 1 | 44 | |||
| AK048630.1 | - | 1 | 42 | |||
| BC094545.1 | - | 24 | ||||