siRNA Details
| Gene Symbol: Pum2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027670.1 | 3, 4 | 409 | |||
| GenBank mRNA | AK122225.1 | - | 3, 4 | 295 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pum2| Gene Symbol: Pum2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027670.1 | 3, 4 | 409 | |||
| GenBank mRNA | AK122225.1 | - | 3, 4 | 295 | ||