siRNA Details
| Gene Symbol: Rreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033218.1 | 4 | 1035 | |||
| NR_033615.1 | 4 | 852 | ||||
| GenBank mRNA | AK047047.1 | - | 4 | 832 | ||
| AK047251.1 | - | 4 | 832 | |||
| AK147260.1 | - | 4 | 1036 | |||
| AK147653.1 | - | 4 | 1036 | |||

Rreb1
Genomic Map