siRNA Details
| Gene Symbol: Gm13749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027824.1 | 2 | 1016 | |||
| GenBank mRNA | AK081719.2 | - | 2 | 1018 | ||
| BC147571.1 | - | 2 | 987 | 9056561 | ||
| BC147573.1 | - | 2 | 987 | 9056563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13749| Gene Symbol: Gm13749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027824.1 | 2 | 1016 | |||
| GenBank mRNA | AK081719.2 | - | 2 | 1018 | ||
| BC147571.1 | - | 2 | 987 | 9056561 | ||
| BC147573.1 | - | 2 | 987 | 9056563 | ||