siRNA Details
| Gene Symbol: Tspy-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027507.1 | 4 | 299 | |||
| GenBank mRNA | AJ001378.1 | - | 4 | 301 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB013038.2 | - | 3 | 628 | Chr. Y - 1055764 - 1057642 [-] on Build GRCm38 | |

Tspy-ps
Genomic Map