siRNA Details
| Gene Symbol: 1700047M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015458.1 | 2 | 481 | |||
| GenBank mRNA | AK006718.1 | - | 2 | 265 | ||
| AK019698.1 | - | 2 | 260 | |||
| AK047804.1 | - | 2242 | ||||
| BC048591.1 | - | 2 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700047M11Rik| Gene Symbol: 1700047M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015458.1 | 2 | 481 | |||
| GenBank mRNA | AK006718.1 | - | 2 | 265 | ||
| AK019698.1 | - | 2 | 260 | |||
| AK047804.1 | - | 2242 | ||||
| BC048591.1 | - | 2 | 481 | |||