siRNA Details
| Gene Symbol: 5033406O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029464.1 | 3 | 537 | |||
| GenBank mRNA | AK017159.1 | - | 3 | 497 | ||
| AK020272.1 | - | 3 | 264 | |||
| AK079861.1 | - | 3 | 531 | |||
| AK134157.1 | - | 3 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5033406O09Rik| Gene Symbol: 5033406O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029464.1 | 3 | 537 | |||
| GenBank mRNA | AK017159.1 | - | 3 | 497 | ||
| AK020272.1 | - | 3 | 264 | |||
| AK079861.1 | - | 3 | 531 | |||
| AK134157.1 | - | 3 | 471 | |||