siRNA Details
| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 2003 | |||
| NR_024422.1 | 2 | 1571 | ||||
| NR_110299.1 | 2 | 1567 | ||||
| NR_110300.1 | 2 | 1339 | ||||
| NR_110301.1 | 2 | 1456 | ||||
| NR_110302.1 | 2 | 1389 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF503-AS2| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 2003 | |||
| NR_024422.1 | 2 | 1571 | ||||
| NR_110299.1 | 2 | 1567 | ||||
| NR_110300.1 | 2 | 1339 | ||||
| NR_110301.1 | 2 | 1456 | ||||
| NR_110302.1 | 2 | 1389 | ||||