siRNA Details
| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 2208 | |||
| NR_024422.1 | 2 | 1776 | ||||
| NR_110299.1 | 2 | 1772 | ||||
| NR_110300.1 | 2 | 1544 | ||||
| NR_110301.1 | 2 | 1661 | ||||
| NR_110302.1 | 2 | 1594 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF503-AS2| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 2208 | |||
| NR_024422.1 | 2 | 1776 | ||||
| NR_110299.1 | 2 | 1772 | ||||
| NR_110300.1 | 2 | 1544 | ||||
| NR_110301.1 | 2 | 1661 | ||||
| NR_110302.1 | 2 | 1594 | ||||