siRNA Details
| Gene Symbol: ZSCAN12P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024063.2 | 2 | 2993 | |||
| GenBank mRNA | AK127188.1 | - | 1 | 515 | ||
| BC040524.1 | - | 2 | 2649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSCAN12P1| Gene Symbol: ZSCAN12P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024063.2 | 2 | 2993 | |||
| GenBank mRNA | AK127188.1 | - | 1 | 515 | ||
| BC040524.1 | - | 2 | 2649 | |||