This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINC00029 SIRNA Assays ›
Gene Symbol:
LINC00029
Gene Name:
long intergenic non-protein coding RNA 29
Gene Aliases:
C20orf51, NCRNA00029
Chromosome Location:
Chr.20: 63034217 - 63037028 on Build GRCh38
UniGene ID:
Hs.187773
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n259428

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00029
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028295.1 1 325
GenBank mRNA AL117533.1 - 1 325

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-cdc4cf686-5rvzh:80/100.66.79.50:80.
git-commit: 18f7f941bc72f70e8ff4e0be257f7363f2314623
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.0-Offline