siRNA Details
| Gene Symbol: LINC00029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028295.1 | 1 | 79 | |||
| GenBank mRNA | AL117533.1 | - | 1 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00029| Gene Symbol: LINC00029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028295.1 | 1 | 79 | |||
| GenBank mRNA | AL117533.1 | - | 1 | 79 | ||