siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 2 | 81 | |||
| GenBank mRNA | BC038721.1 | - | 2 | 84 | ||
| BC048119.1 | - | 2 | 82 | |||
| BC104172.1 | - | 2 | 33 | |||
| BC115360.1 | - | 2 | 33 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00879| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 2 | 81 | |||
| GenBank mRNA | BC038721.1 | - | 2 | 84 | ||
| BC048119.1 | - | 2 | 82 | |||
| BC104172.1 | - | 2 | 33 | |||
| BC115360.1 | - | 2 | 33 | |||