This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PART1 SIRNA Assays ›
Gene Symbol:
PART1
Gene Name:
prostate androgen-regulated transcript 1 (non-protein coding)
Gene Aliases:
NCRNA00206
Chromosome Location:
Chr.5: 60487713 - 60547657 on Build GRCh38
UniGene ID:
Hs.146312
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n260057

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PART1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024617.1 1 469
NR_028508.1 1 250
NR_028509.1 1 250
GenBank mRNA AF163475.1 - 1 250
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA930113.1 - 1 44 Chr. 5 - 60488140 - 60546164 [+] on Build GRCh38
GenBankmRNA DA296238.1 - 1 205 Chr. 5 - 60487977 - 60529508 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-qjpw4:80/100.66.76.224:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0