siRNA Details
| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024617.1 | 1 | 380 | |||
| NR_028508.1 | 1 | 161 | ||||
| NR_028509.1 | 1 | 161 | ||||
| GenBank mRNA | AF163475.1 | - | 1 | 161 | ||
| AK312071.1 | - | 1 | 383 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA296238.1 | - | 1 | 116 | Chr. 5 - 60487977 - 60529508 [+] on Build GRCh38 | |

PART1
Genomic Map