This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAM5 SIRNA Assays ›
Gene Symbol:
ADAM5
Gene Name:
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases:
ADAM5P, TMDCII
Chromosome Location:
Chr.8: 39314663 - 39417226 on Build GRCh38
UniGene ID:
Hs.369765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n260954

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ADAM5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001448.1 8, 9 778
GenBank mRNA AJ132821.1 - 7, 8 712
AJ132822.1 - 7, 8 594
AJ132823.1 - 6, 7 528
BC047448.1 - 8, 9 912
BC067864.1 - 6, 7 571
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI560028.1 - 6, 7 575 Chr. 8 - 39314644 - 39375865 [+] on Build GRCh38
GenBankmRNA BI829318.1 - 6, 7 665 Chr. 8 - 39314656 - 39375761 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
metalloendopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline