This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SNORA64 SIRNA Assays ›
Gene Symbol:
SNORA64
Gene Name:
small nucleolar RNA, H/ACA box 64
Gene Aliases:
RNU64, U64
Chromosome Location:
Chr.16: 1962973 - 1963106 on Build GRCh38
UniGene ID:
Hs.745496
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n261364

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNORA64
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002326.1 1 78
GenBank mRNA Y11158.1 - 1 78
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE936551.1 - 1 230 Chr. 16 - 1962942 - 1963244 [-] on Build GRCh38
GenBankmRNA CN421741.1 - 3 494 Chr. 16 - 1962775 - 1964624 [-] on Build GRCh38
GenBankmRNA DR157078.1 - 3 504 Chr. 16 - 1962855 - 1964631 [-] on Build GRCh38
GenBankmRNA CN421747.1 - 1 230 Chr. 16 - 1962629 - 1963258 [-] on Build GRCh38
GenBankmRNA BM563662.1 - 523 Chr. - - [-] on Build
GenBankmRNA CN405534.1 - 1 78 Chr. 16 - 1962214 - 1963096 [-] on Build GRCh38
GenBankmRNA CN421735.1 - 3 473 Chr. 16 - 1962947 - 1964602 [-] on Build GRCh38
GenBankmRNA BE263765.1 - 3 486 Chr. 16 - 1962820 - 1964616 [-] on Build GRCh38
GenBankmRNA CN405671.1 - 2 292 Chr. 16 - 1962497 - 1964333 [-] on Build GRCh38
GenBankmRNA CN421739.1 - 3 483 Chr. 16 - 1962877 - 1964613 [-] on Build GRCh38
GenBankmRNA CX781018.1 - 3 451 Chr. 16 - 1962579 - 1964581 [-] on Build GRCh38
GenBankmRNA T68031.1 - 138 Chr. - - [-] on Build
GenBankmRNA CR996341.1 - 3 506 Chr. 16 - 1962882 - 1964631 [-] on Build GRCh38
GenBankmRNA BI910102.1 - 3 500 Chr. 16 - 1962712 - 1964631 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-kfcqn:80/100.66.75.174:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0