siRNA Details
| Gene Symbol: FTH1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002201.1 | 1 | 748 | |||
| GenBank mRNA | BC073777.1 | - | 1 | 759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FTH1P3| Gene Symbol: FTH1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002201.1 | 1 | 748 | |||
| GenBank mRNA | BC073777.1 | - | 1 | 759 | ||