This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KLRA1P SIRNA Assays ›
Gene Symbol:
KLRA1P
Gene Name:
killer cell lectin like receptor A1, pseudogene
Gene Aliases:
KLRA1, KLRAP1, LY49L, Ly-49L, Ly49
Chromosome Location:
Chr.12: 10588478 - 10599835 on Build GRCh38
UniGene ID:
Hs.159297
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n261897

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLRA1P
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028045.1 4 606
GenBank mRNA AF047445.1 - 4 336
AK290077.1 - 3 307
AK296103.1 - 4 343
AK304677.1 - 3 302
AY334570.1 - 2 150
BC069672.1 - 4 309 7262588
BC069697.1 - 4 309 7262600
BC069734.1 - 4 309 7262612
BC101471.1 - 4 323 8068977
BC101473.1 - 4 326 8068979

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-jfh8m:80/100.66.78.238:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0